Learn More
Trajan™ Replacement Needles for Thermo Scientific GC Syringes
Cone tip style. Trajan Scientific and Medical™ Replacement Needles for Thermo Scientific GC Autosampler Syringes are not gas tight.
$90.00 - $111.00
Specifications
Needle Point Style Cone
View More Specs
Products 2
Catalog Number Mfr. No. Needle Gauge Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Needle Gauge Product Type Price Quantity & Availability  
03-350-127
View Documents
Trajan™
037787
23 G GC Autosampler Syringe Replacement Needle
Pack of 2 for $90.00
Add to Cart
03-350-124
View Documents
Trajan™
033770
26 G GC Autosampler Syringe Replacement Needle
Each for $111.00
Add to Cart
Description
Description
  • Needle length is the exposed length of needle
  • Fits Thermo Scientific™ syringes for AS3000, AS2000 and TriPlus RSH Autosamplers
Specifications
Specifications
Cone
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.