Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non-specific sample interaction and increased analyte recovery.
Supplier:  Trajan™ 002790
 View more versions of this product
Catalog No. SG002790
Explore all special offers
Add to Cart
Description
Description
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
Specifications
Gas/Liquid chromatography
662°F
10R-RTC/RSH-GT-MS1
10 μL
Removable
Autosampler Syringe
350°C
Ea.
Thermo RSH and CTC RTC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.