Learn More
Lonza Walkersville SG Cell Line 96-well Nucleofector Kit
Supplier: Lonza Walkersville V4SC3096
The SG Cell Line 96-well Nucleofector Kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D-Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:2.25 mL SG Cell Line Nucleofector Solution0.5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)1 x 96-well Nucleocuvette Plate (20 μl)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.