Learn More
Lonza Walkersville SF Cell Line 4D-Nucleofector X Kit L
Supplier: Lonza Walkersville V4XC2012
The SF Cell Line 4D-Nucleofector X Kit L is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput Applications Content:2 x 0.675 mL SF Cell Line Nucleofector Solution2 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)12 x 100 µL Nucleocuvette Vessel12 x Single use pipettes
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.