Learn More
Lonza Walkersville SF Cell Line 4D-Nucleofector X Kit L, 24 reactions, 100uL
Supplier: Lonza Walkersville V4XC2024
The SF Cell Line 4D-Nucleofector X Kit L is one of three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit. This kit is optimal for transfecting the following cell types: 293-F (FreeStyle), 32D, A-20, A2058, A375, A431, A549, AGS, ARPE-19, BetaTC/6, BxPC-3, C6, CCRF-CEM, CHO-K1, CHO-S (Invitrogen), Cos-1, EL4, FDC-P1, H9c2 (2-1), HEK293, Hel9217, HepG2, High 5, HL-60, HMC-1-8, HT-1080, HT-29, Hut-78, HUV-EC-C, IMR-32, J774A.1, K-562, KG-1, L-428, LNCaP, MDA-MB-453, MDBK, Molt-4, MV-4-11, Nalm-6, NB-4, NCI-H1299 [H1299], Neuro-2a [N2a], NG108-15, NS0, P19, P815, PC-12, PC-3, Raw 264.7, RBL-1, RBL-2H3, Reh, Rin m5F, RPMI 8226, S2, S49, SaOS, SF9, SH-SY5Y, SK-BR-3, SK-N-AS, SK-N-MC, SK-N-SH, SK-OV-3, Sp2/0-Ag14, SW480, T2, T84, TF-1, U266B1, U-937, and Vero.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.