Learn More
KRISHGEN BIOSYSTEMS Endonuclease Serratia marcescens ELISA for quantitative determination of endonuclease in samples from downstream processing where endonuclease is used as a process or purification aid.
Supplier:  KRISHGEN BIOSYSTEMS KBBA36
Encompass_Preferred
Pack Size: 1 x 96 wells. Assay Range: 0 - 256 ng/ml, Sensitivity: 0.1 ng/ml. Introduction: Serratia marcescens nuclease originates from Gram negative bacteria S. marcescens and heads a family of homological non-specific nucleases that are widely spread in the world. Serratia nuclease is most studied one and it is capable to cleavage both RNA and DNA in either single or double stranded form. Principle: The Endonuclease Serratia marcescens (FastNuclease) ELISA is a competitive immunoassay for the determination of Serratia endonuclease in sample. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747162
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.