Learn More
Abnova™ SERPINB6/CEN6p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
$2382.00
Specifications
Location | 6p25, 6p12.1 |
---|---|
Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Product Type | SERPINB6/CEN6p FISH Probe |
Probe | SERPINB6, CEN6p |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-024-943
|
Abnova™
FG0046 |
200 uL |
Each of 1 for $2,382.00
|
|
|||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
6p25, 6p12.1 | |
Fluorescent In Situ Hybridization (Cell) | |
SERPINB6, CEN6p | |
Texas Red, FITC |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
SERPINB6/CEN6p FISH Probe | |
Store at 4°C in the dark. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.