Learn More
BINDER™ Series ED Classic.Line Drying and Heating Chambers with Gravity Convection
BINDER™ Series ED Classic.Line Drying and Heating Chambers ensure fast and uniform drying.
Supplier: BINDER™ 90100193
Description
- APT.line™ preheating chamber technology
- Gravity convection
- Adjustable exhaust air flap
- Controller with timer functions
- 2 chrome-plated racks
- Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
Specifications
With RS 442 | |
5°C above room temperature to 300°C | |
1 door | |
ED023UL-120V-I | |
300 x 222 x 330 mm | |
120 V, 60 Hz, 12.5 A |
20 L | |
Drying and Heating Chamber | |
12 kg | |
2 std. / 4 max. | |
520 x 435 x 495 mm |