Learn More
Electron Microscopy Sciences Sequenza™ Slide Rack
Supplier: Electron Microscopy Sciences 7140701
Replacement parts for Sequenza, a system designed for immunolabeling of sections on glass slides. Ideal for both benchtop techniques and microwave-assisted applications. This system maximizes the results produced up to 50 slides per run, maintaining both consistency and quality. The Slide Rack holds 10 glass slides, with a coverplate forming a capillary gap between the slide and the plate. A hopper easily dispenses the reagent into the gap formed between it and the coverplate. The Slide Rack holds 10 glass slides.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.