Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ SemperForce™ Black Nitrile Gloves

Catalog No. 1904858B Shop All Sempermed USA Inc. Products
Click to view available options
Size:
This item is not returnable. View return policy
This item is not returnable. View return policy

Black nitrile gloves feature textured fingertips to ensure secure gripping capabilities for demanding work environments.

  • Enhanced dexterity and gripping capabilities
  • Hides stains caused by grease, dirt, dyes and ink
  • ESD tested and approved
  • Ideal for industrial, electronics, automotive applications

Specifications

Certifications/Compliance

FDA, ESD tested and approved

Applications

Industrial

Color

Black

Disposable

Single-use

Exterior Finish

Partially Textured

Latex Free

Yes

Material

Nitrile

Powdered

Powder Free

Size

Medium

Thickness (English)

4.5 mil

Thickness (Metric)

0.11 mm

Model

BKNF103

Length (English)

9 in.

Length (Metric)

23 cm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.