Learn More
Electron Microscopy Sciences SEM Pin Stub Adapter, 12.5 mm
Supplier:  Electron Microscopy Sciences EMS062
Encompass_Preferred
A simple to use adapter which will convert Hitachi, JEOL or ISI microscopes to use 1/8" pin stubs or Cambridge stubs. Made of aluminum with spring retainers which will give a good connection.
Catalog No. 50-193-0524
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.