Learn More
Electron Microscopy Sciences Cylinder Mount SEM 3/8" head, 3/16" height, Aluminum
Supplier: Electron Microscopy Sciences 75360
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 3/8"
Pin height: 3/16"