Learn More
Electron Microscopy Sciences SEM Hitachi S-450 M4 Mount 3-Division 5/8", 1/4" height, Aluminum
Supplier: Electron Microscopy Sciences 75600D
Hitachi S-450 SEM mounts with M4 tapered hole and female thread. With 3 divisions. Made from ultra-pure aluminum with standard lathe finish.
Head: 5/8"
Height: 1/4"