Learn More
Electron Microscopy Sciences Scanning Electron Microscope Finder Grid, Copper
Supplier: Electron Microscopy Sciences 80100CU
Finder grids for SEM; a valuable tool for analytical studies. The grids are available in 10 mm diameter and they can be placed directly on the stub surface and used to identify the area of interest in which the studies are performed.