Learn More
Electron Microscopy Sciences SEM Cylinder Mount 45° Angle 1" Surface, Aluminum
Supplier: Electron Microscopy Sciences 7524160
Scanning electron microscope specimen mount with 45 degree angle. Made from lathe finished ultra-pure aluminum.
Head: 1"
Height: 13/16"