Learn More
Eurofins Discoverx Secretin, 250 μg. (92-1181)
Supplier:  Eurofins Discoverx 921181
Encompass_Preferred
Secretin is a 27 amino acid GI peptide hormone acting as an agonist for SCTR. It is responsible for regulation of water homeostasis, secretions of the stomach and pancreas, and pH of the duodenum. May also play a role in osmoregulation of the hypothalamus, pituitary, and kidney. Provided as 250 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9709
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.