Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 20mm Diameter, Medium Porosity, Tube O.D./25mm
SDP
Supplier:  Chemglass Life Sciences CG20206
Encompass_Preferred
Glassblowers Fritted Tube, Straight, Frit 20mm Diameter, Medium Porosity, Tube O.D./25mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.
Catalog No. CG20206
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.