Learn More
Chemglass Life Sciences Crimp Seal, Silver, PTFE/Silicone/PTFE
SDP
Supplier:  Chemglass Life Sciences CV34000008
Encompass_Preferred
Crimp Seal, Silver, PTFE/Silicone/PTFE. Crimp Seal Selection Guide:PTFE/Red Rubber -Medium durometer red rubber with a .015 layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.PTFE/Silicone -Pure Silicone laminated with .005 of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities.PTFE/Silicone/PTFE -Medium durometer silicone with .003 silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.Viton -Recommended for chlorinated solvents. Excellent chemical resistance.Polypropylene -Not resealable. For one time use only. Easily penetrated by syringes and chemical resistant.
Catalog No. 50-230-969
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.