Learn More
MTC Bio Screw Cap Microtube, 2.0mL, capless, no printing (no graduations/marking area) self-standing, non-sterile, 1000/bag
Supplier:  MTC Bio C3172
Encompass_Preferred
The newest addition to the ClearSeal™ line is made of 100% natural/colorless polypropylene with no markings, dyes or metallic content and they conform with FDA 21CFR. In response to popular demand for diagnostic sampling and reagent packaging applications, color coded caps are also available. The silicone O-ring ensures a leakproof seal throughout the temperature range of -90°C to +121° C. All ClearSeal™ tubes are RNase, DNase, pyrogen/endotoxin free and may be centrifuged at up to 22,000xg. Dimensions 13.1 x 46.8cm.
Catalog No. 50-226-8460
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.