Learn More
MTC Bio Culture Tube, 5mL, 12 x 75mm, PS, w/ attached 2-position screw-cap, printed graduations, sterile, 20 bags of 25 tubes, 500/case
Supplier: MTC Bio T8741
MTC Bio’s new DuoClick™ tubes feature a unique, patent pending screw closure system. DuoClick tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing. Just rotate the cap 1/4 turn to the first click to secure the cap in the aerobic position. Or, rotate it another 1/2 turn past the second click for a leakproof, airtight seal for anaerobic culturing. DuoClick tubes are available gamma sterilized, with screw caps attached, and in our exclusive ready-to-use rack packaging. Tubes feature clear printed graduations, and are sterilized by electron beam. RNase, DNase, pyrogen, & endotoxin free.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.