Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
Ideal for peak identification work
Supplier: Restek Corporation 34497
Description
- CGA 165 connection and can deliver large volumes of sample
- Scotty™ (Air Liquide) 48
- Contents: 48L
- Pressure: 300 psig (21 bar)
- Outlet Fitting: CGA 165
- Weight: 1.75 lb. (0.8kg)
- Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
- DOT Specifications: 39 NRC
All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements
Specifications
Air Standard | |
10.2 cm | |
1 ppm | |
300 psi | |
DOT 39 NRC | |
Vinyl Chloride in Nitrogen | |
12.68 gal. |
41 cm | |
48 L | |
0.8 kg | |
1.75 lb. | |
Ideal for peak identification work | |
16.25 in. | |
4 in. |
Safety and Handling
ShelfLife : 2 Years
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction