Learn More
Electron Microscopy Sciences Scott's Tap Water Solution 1 LT
Supplier:  Electron Microscopy Sciences 2607006
Encompass_Preferred
Magnesium sulfate buffered with sodium bicarbonate solution is a prepared, ready-to-use, high quality solution that saves time and effort.
Catalog No. 50-285-35
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.