Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scotch™ Glass Cloth Electrical Tape
Scotch™ Glass Cloth Electrical Tape is premium grade and edge tear resistant.
Supplier: 3M™ 7000005818
Description
- Printable
- Non corrosive adhesive
- Offers solvent resistant protection
- Flame retardant and suitable for 600V rated, dry location application motors and transformers.
- Thermosetting, silicon adhesive provides an increased bond at high temperatures
- Glass cloth backing optimally resists resist shrinking and burning
- Used as a coil cover, anchor, banding, core, layer and crossover insulation
- Printable and provides high mechanical strength and excellent performance by withstanding high temperatures
- Withstands a temperature range of 32 to 392°F (0 to 200°C)
- UL Listed, Mil-I-19166C, CSA Certified and RoHS 2011/65/EU Compliant
Specifications
White | |
Glass Cloth Tape | |
69-3/4″X66′ |
Silicone | |
Communications, Consumer, Design and Construction, Energy, Healthcare, Manufacturing, Mining, Oil and Gas | |
66 ft. x 3/4 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction