Learn More
Electron Microscopy Sciences Scotch™ 666 Double Sided Tape 72 YD
Supplier:  Electron Microscopy Sciences 77116
Encompass_Preferred
Non-conductive transparent tape with adhesive on both sides. With white paper liner, which is slit in the center to facilitate its removal. This produces good adhesion and cleanliness. 3" core.
Catalog No. 50-285-31
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.