Learn More
Electron Microscopy Sciences SEM Scintillator YAP 18.0x1 mm
Supplier: Electron Microscopy Sciences 82084
YAP (Yttrium Aluminum Perovskite activated by Ce3+) shares the same advantages as YAG but is more efficient in terms of light output than YAG crystals. As well, the emission spectrum peaks at about 378nm, and this corresponds closely to the maximum sensitivity of the S11 photocathode which in general is used in most scanning electron microscopes. (There would be more improvement in signal by using a YAP crystal rather than YAG in these microscopes). The decay time of YAP (40ns) is faster than YAG (80ns) so its overall performance is superior to YAG.
YAP crystals should be coated with 50nm of aluminum prior to use. Note: P-47 is usually in-stock. YAG and YAP delivery time is 6-8 weeks. Use with Camscan, Balscan, Camb scanning electron microscopes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.