Learn More
ILEX LIFE SCIENCES LLC SCHNEIDERS P04-90599 20X500ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0490599X20
Encompass_Preferred
PAN-Biotech Schneider’s Drosophila Medium, w/o: L-Glutamine, w/o: L-Methionine, w: 0.40 g/L NaHCO3 (20 x 500 ml). BICARBONATE: With 0.4 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. L-METHIONINE: Without L-Methionine. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Originally developed for the culture of Drosophila cells, this medium is also suitable for the culture of other dipteran cell lines.
Catalog No. 50-273-3758
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.