Learn More
Crescent Chemical Co Inc SCHNEIDER S DROSOPHILA POWDER
Supplier: Crescent Chemical Co Inc SE47521.04
Schneider's Drosophila powder medium revised 24,89 g/L Without sodium bicarbonate, without calcium chloride. Size - 10L Storage Conditions - +2 °C TO +8 °C Catalog Number - 47521.04