Learn More
Andwin Scientific SCALPEL NO11 FEATHER DISP 20PK
Supplier: Andwin Scientific FH/CX7281A
The Feather Safeshield scalpels are ideal for delicate precise procedures with its heat-treated stainless steal material. This product features and comfort and protection when utilized. The scalpels minimize injuries and transmitted disease that are obtained from contaminated blades. These also have a clear protective, retractable shield and a color coded button on the protective shield which helps in the identification of the blade size. This product is in style #11 and is sold in a pack of 20.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.