Learn More
Sarstedt Inc Screw Cap Microtube, 1.3mL, 1000/Case
Supplier: Sarstedt Inc 41.1393.105
Microtube, Prepared, with screw cap closure, Plasma, Lithium Heparin, 1.3mL, 1000cs, Length: 45mm, Diameter: 10.8mm, Closure contains integral O-ring, Skirted, conical base, and labels