Learn More
Enzo Life Sciences SC-51089 (25 mg). CAS: 146033-02-5
SDP
Supplier:  Enzo Life Sciences BMLRA1130025
Encompass_Preferred
A potent and selective prostaglandin E2 (EP1) antagonist. Displays in vivo analgesic activity in the rat. Does not block PGE1-induced hyperalgesia. Purity: ≥98% (HPLC). Solubility: Soluble in water or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1719
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.