Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit
Supplier:  ACROBiosystems RASA01096TESTS
Encompass_Preferred
Kits;96TESTS;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Nucleocapsid Protein. The kit consists of Anti-SARS-CoV-2 Nucleocapsid Antibody, an SARS-CoV-2 Nucleocapsid Protein as Control, an biotin-Anti-SARS-CoV-2 Nucleocapsid Antibody and HRP-Streptavidin. ;Detection of Monoclonal SARS-CoV-2 Nucleocapsid Protein titer by sandwich-ELISA Assay.
Immobilized Anti-SARS-CoV-2 Nucleocapsid Protein Antibody at 4 μg/mL (100 μL/well) can bind SARS-CoV-2 Nucleocapsid Protein and Biotin-Anti-SARS-CoV-2 Nucleocapsid Antibody. Detection was performed using Streptavidin-HRP with sensitivity of 12.5 pg/mL (QC tested).
Catalog No. 50-210-7444
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.