Learn More
Cole-Parmer Fitting, 304 Stainless Steel, Sanitary Clamp, 2"
Supplier:  Cole-Parmer 5011844
Encompass_Preferred
Use the sanitary clamps with sanitary fittings to create a leak-proof connection. These easy-to-use clamps are made from 304 stainless steel to provides good chemical resistance.
Catalog No. 50-255-7744
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.