Learn More
Chemplex Industries SAMPLECUPS 30.7MM DIA.SNAPPOST
Supplier: Chemplex Industries 1730SE
CHEMPLEX XRF SAMPLE CUPS, 30.7 Dia., mm, Snap-Post Venting, Snap-On Rings have integrated thin-film trimmer