Learn More
Chemplex Industries SAMPLE CUPS 30.7MM DIA. DBL OP
Supplier: Chemplex Industries 1330SE
CHEMPLEX XRF SAMPLE CUPS, 30.7 Dia., mm, Double open ends, Snap-On Rings have integrated thinfilm trimmer