Learn More
Thermo Scientific™ Samco™ Transfer Pipettes
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Supplier: Thermo Scientific™ 273
Description
Prevent cross-contamination with Thermo ScientificTM SamcoTM General-Purpose Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface reduces loss of cells and valuable proteins due to binding. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for:Tissue culture and applications, routing sampling of serum and plasma, blood culture, urine analysis
Your input is important to us. Please complete this form to provide feedback related to the content on this product.