Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDC SafeGuard™ Direct Thermal Slide Labels
Direct thermal chemical resistant slide labels are 4-across and perforated between each row.
Supplier: PDC TD141516
Description
- Protect Patient Data to Prevent Medical Errors
- Direct thermal, chemical-resistant slide labels - ideal for use in histology testing
- Prevent barcode degradation and adhesive oozing that can lead to misidentification
- Highly resistant to harsh chemicals including xylene, hexane, staining reagents, alcohol, acetone, and more
- Quick and easy printing with a direct thermal printer, with no need for an overlay or etching system
Specifications
Permanent (Rubber-based) | |
White | |
Die-cut | |
Single-side Printable | |
Direct Thermal Printable Label | |
0.94 x 0.94 in. | |
0.94 in. |
None | |
Gloss | |
Plastic | |
Direct Thermal | |
Non-sterile | |
0.94 in. | |
2.38 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction