Learn More
BioVendor, LLC S2000 Denatured Collagen Stnd
Supplier: BioVendor, LLC S2010
Denatured collagen standard for the Biocolor S2000 Sircol Insoluble Collagen (Bovine) colorimetric assay. 1mg/ml (3x5ml). A sterile solution of cold, water-soluble denatured collagen from bovine skin, stored in 0.1M acetic acid within a sealed vial. Once opened, store at 4 degrees C, discard if the solution becomes turbid.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.