Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt
Group I agonist
$313.00 - $1308.00
Specifications
CAS | 7381-67-1 |
---|---|
Solubility | Soluble to 100 mM in water |
Molecular Formula | C3H6NO5S2Na |
Formula Weight | Observed MW: 223.19 |
Description
EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.Specifications
7381-67-1 | |
Soluble to 100 mM in water | |
C3H6NO5S2Na | |
Observed MW: 223.19 |
Safety and Handling
Recommended Storage : Desiccate at -20°C
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title