Learn More
Sarstedt Inc S-MONO 1.2ML LI HEP 8X66/PK50
Supplier:  Sarstedt Inc 06.1666.100
Encompass_Preferred
S-Monovette Venous Blood Collection Tube 1.2mL Lithium Heparin 8x66mm Sterile
Catalog No. NC9066444
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.