Learn More
Enzo Life Sciences S-Farnesyl-L-cysteine methyl ester (25mg). CAS: 125741-64-2
Supplier: Enzo Life Sciences BMLG2010025
Stimulates the multidrug resistance transporter ATPase activity (4-5-fold at 10-20 µM) and competes for drug binding. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) and ethanol (25 mg/ml). Long Term Storage: -20°C.