Learn More
Enzo Life Sciences Rucaparib phosphate (25mg). CAS: 459868-92-9
SDP
Supplier:  Enzo Life Sciences ENZCHM2360025
Encompass_Preferred
Rucaparib is a poly (ADP ribose) polymerase (PARP) inhibitor. PARP is a DNA damage-activated nuclear enzyme that has a key signaling role in the base excision repair pathway. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3652
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.