Learn More
Andwin Scientific Pipette Tips RT LTS 5000 uL 192A/8, 192/pk
SDP
Supplier:  Andwin Scientific 30389256
Encompass
Racked tips, 5000 uL max. volume, for Rainin pipettes with LTS LiteTouch, 192 tips in 8 racks of 24.
Catalog No. NC1782795
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.