Learn More
STEMCELL Technologies RS-1
SDP
Supplier:  STEMCELL Technologies 74092
Encompass_Preferred
HDR pathway enhancer; Enhances RAD51 - 10 Mg
Catalog No. 50-274-0757
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.