Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ RPMI 1640 Medium
General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes
Supplier: MP Biomedicals™ 091646754
Specifications
0° to 5°C | |
Sterile | |
6.8 to 7.4 |
RPMI 1640 Medium | |
No Glutamine, No Phenol Red | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction