Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
Supplier:  Thermo Scientific™ 75003865
 View more versions of this product
Catalog No. 75-003-865
Description
Description
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
Specifications
HAEMAFlex 12/16 swinging bucket rotors
16 x 500 mL
Higher capacity, 12/16 Swing Bucket Rotors
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.