Learn More
Hettich Centrifuges ROTANTA460,(196)13(48)16,Pk
SDP
Supplier:  Hettich Centrifuges 460BLOODHC
Encompass_Preferred
Hettich ROTANTA 460 BLOOD TUBE PACKAGE HIGH CAPACITY, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 13 x 75/100 mm, (4) Adapters 16 x 75/100, (4) Bio-containment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7580
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.