Learn More
Enzo Life Sciences ROS-ID Superoxide detection kit (1Kit)
Supplier: Enzo Life Sciences ENZ51012
Enzo Life Sciences’ ROS-ID Superoxide detection kit is designed to directly monitor real time superoxide production in live cells using fluorescence microscopy and/or flow cytometry. A major component of the kit, Superoxide Detection Reagent (Orange), is a cell-permeable probe that reacts specifically with superoxide, generating an orange fluorescent product. The kit is not designed to detect reactive peroxide, hydroxyl, peroxynitrite, chlorine or bromine species, as the fluorescent probe included is relatively insensitive to these analytes. Upon staining, the fluorescent product generated can be visualized using a wide-field fluorescence microscope equipped with a standard orange (e.g., 550/620 nm) fluorescent cube, or cytometrically using any flow cytometer equipped with a blue (488 nm) laser. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.