Learn More
Enzo Life Sciences ROS-ID Hypoxia/Oxidative stress detection kit (125tests)
SDP
Supplier:  Enzo Life Sciences ENZ510420125
Encompass_Preferred
Enzo Life Sciences’ ROS-ID Hypoxia/Oxidative stress detection kit is designed for functional detection of hypoxia and oxidative stress levels in live cells (both suspension and adherent) using fluorescent microscopy or flow cytometry. This kit includes fluorogenic probes for hypoxia (red) and for oxidative stress levels (green) as two major components. The Hypoxia (Red) dye takes advantage of the nitroreductase activity present in hypoxic cells by converting the Nitro group to hydroxylamine (NHOH) and amino (NH2) and releasing the fluorescent probe. The generated fluorescent products can be visualized using a wide-field fluorescence microscope equipped with standard fluorescein (490/525 nm) and Texas Red (596/670 nm) filters, confocal microscopy, or cytometrically using any flow cytometer equipped with a blue (488 nm) laser. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
Catalog No. 50-201-2601
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.