Learn More
Electron Microscopy Sciences Acrylic Rodent Brain Matrix Rat, 300-600g, Coronal
Supplier: Electron Microscopy Sciences 69083C
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Size: 300-600g
Type: Coronal
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.