Learn More
Sklar Instruments Forceps, Merit Rochester-Pean Hemostatic Forceps, Sklar 8 in., Curved, Serrated, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 97498
Encompass_Preferred
Forceps, Merit Rochester-Pean Hemostatic Forceps, Sklar, 8 in., Curved, Serrated, Stainless Steel, EA. Merit Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is curvedt with full, horizontally serrated jaws, and a length of 8 inches.
Catalog No. 50-112-6634
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.